Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Revolution™ Harnesses

Catalog No. 141305000 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect Buckle Leg Straps
Tongue-Buckle Leg Straps
Material:
DualTech Webbing
Dupont™ Kevlar™ aramid fiber-Nomex™ aramid fibers webbing
DuraFlex Webbing
Type 10 Webbing
Style:
Black with Dupont™ Kevlar™-Nomex™ aramid fibers webbing
Standard Harness
With Removable Belt
With Removable belt, Side D-Rings and Pad
With Side D-rings and pad
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) Material Style
14-130-5000 Quick-Connect Buckle Leg Straps DuraFlex Webbing Standard Harness
19-034-867 Tongue-Buckle Leg Straps DualTech Webbing With Side D-rings and pad
19-130-4080 Quick-Connect Buckle Leg Straps DualTech Webbing Standard Harness
19-130-4104 Quick-Connect Buckle Leg Straps Dupont™ Kevlar™ aramid fiber-Nomex™ aramid fibers webbing Black with Dupont™ Kevlar™-Nomex™ aramid fibers webbing
19-034-869 Quick-Connect Buckle Leg Straps DualTech Webbing With Side D-rings and pad
19-034-868 Tongue-Buckle Leg Straps DualTech Webbing With Removable Belt
17-987-603 Tongue-Buckle Leg Straps Type 10 Webbing With Removable belt, Side D-Rings and Pad
17-987-601 Tongue-Buckle Leg Straps Type 10 Webbing Standard Harness
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 14-130-5000 Supplier Honeywell™ Supplier No. RDFQC/UGN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Design based on key user-identified needs

  • PivotLink™ connection with unique rotary design promotes greater comfort in bending/mobility; also serves as an attachment point for belts, tools and accessories as part of an integrated accessory system
  • Cam buckle provides easy, one-handed adjustment/release and allows simultaneous adjustment of shoulder straps
  • Semi-flexible ErgoArmor™ back shield minimizes impact from sharp/heavy snap hooks and self-retracting lifelines; knitted mesh membrane minimizes moisture absorption and maintains optimal air circulation
  • Stand-up back D-ring simplifies connection
  • DualTech™ webbing makes donning easy with two-sided, contrasting-color webbing and textures
  • High-impact-resistant polymer web finials provide positive gripping surface for easy web adjustment
  • Integrated self-contained label pack encapsulates labels, decreasing damage and loss

Order Info

Webbing tested to 7000 lb. tensile strength. Universal size.

Compliance

Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; CSA Z259.10 requirements.

Specifications

Color Black
Material DuraFlex Webbing
Product Type Revolution Harness
Buckle Style (Leg) Quick-Connect Buckle Leg Straps
For Use With (Application) Construction, Fall Protection
Size Universal
Style Standard Harness
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.