Learn More
Honeywell™ Miller™ Revolution™ Harnesses Accessories: Belt
Catalog No. 191304453
Change view
Click to view available options
Includes:
Five connectors tool (support belt only)
Pad; PivotLink attachment; 5 Accessory connectors
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-130-4453 Pad; PivotLink attachment; 5 Accessory connectors
19-130-4452 Five connectors tool (support belt only)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-4453 Supplier Honeywell™ Supplier No. RIAB2/
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
The PivotLink Connection provides an attachment point for a multitude of accessories and individual tool pouches. The tool belt easily attaches and detaches - distributing weight across the hips and shoulders for a more comfortable fit and greater mobility.
  • Revolution Belts with PivotLink attachments and 5 accessory connectors
  • Innovative design accommodates: pouches, tool holders, cell phones, water bottles and more
  • Universal attachment clips that fit on your existing accessories or tool rig and easily attach/detach
TRUSTED_SUSTAINABILITY
Specifications
Product Type Belt w/pad
Includes Pad; PivotLink attachment; 5 Accessory connectors