Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Step-In and Quick-Connect Harnesses

Catalog No. 19034824A
Change view
Click to view available options
Includes:
Pull-free lanyard rings
Pull-free lanyard rings and sub-pelvic strap
Size:
Small/Medium
Universal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
19-034-824A Pull-free lanyard rings and sub-pelvic strap Universal
19-034-825A Pull-free lanyard rings Universal
19-034-824 Pull-free lanyard rings and sub-pelvic strap Small/Medium
19-034-825 Pull-free lanyard rings Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-034-824A Supplier Honeywell™ Supplier No. 751/UYK

Designed to meet the challenges of harsh working environments.

  • Economical harnesses made of lightweight, durable polyester webbing are easy to put on and take off
  • Friction-buckle shoulder straps and pass-through chest buckle
  • Chest and back straps keep shoulder straps in place and keep the sliding back D-ring from slipping
  • Pull-free lanyard rings and mating buckles in leg straps
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 181.4 kg
Buckle Style (Chest) Mating buckle
Capacity (English) 400 lb.
D-Ring Location Back
Size Universal
Certifications/Compliance CSA; OSHA
For Use With (Application) Confined Space; Construction
Webbing Material Polyester
Product Type Standard non-stretch harnesses
Includes Pull-free lanyard rings and sub-pelvic strap
Style Vest
Buckle Style (Leg) Mating buckle
Color Black
Material Steel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.