Learn More
Honeywell™ Miller™ StretchStop™ Lanyard with SofStop Shock Absorber
Catalog No. 19034896 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Connector Types:
2 Locking snap hooks
Locking Snap Hook and Locking Rebar Hook
Leg Configuration:
Single-Leg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Leg Configuration
19-034-896 Locking Snap Hook and Locking Rebar Hook Single-Leg
19-066-225 2 Locking snap hooks Single-Leg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-896 Supplier Honeywell™ Supplier No. 913RSSZ7/6FTGNC
Combination of integral SofStop Shock Absorber with stretchable lanyard design
Choose from three options:
  • 6-ft. single leg, 2 locking snap hooks
  • ANSI Z359 Compliant, Two legs, locking snap hook and 2 locking rebar hooks (2.5 in.)
  • ANSI Z359 Compliant, 6-ft. single leg, 1 locking snap hook and 1 locking rebar hook (2.5 in.)
Compliance
ANSI Z359 Compliant.
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Product Type Web Lanyard, ANSI Z359 Compliant, single leg
Leg Configuration Single-Leg
Length (English) 6 ft.
Connector Types Locking Snap Hook and Locking Rebar Hook