Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millex™-GP Syringe Filter, PES, 0.45 μm

Catalog No. SLHP033NB Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric) Filter:
13 mm
33 mm
Quantity:
100/Pk.
1000/Pk.
250/Pk.
50/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Filter Quantity
SLHP033NB 33 mm 250/Pk.
SLHPX13NL 13 mm 100/Pk.
SLHP033NS 33 mm 50/Pk.
SLHP033NK 33 mm 1000/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SLHP033NB Supplier MilliporeSigma™ Supplier No. SLHP033NB
Add to Cart
Add to Cart

Millex™-GP PES Syringe Filter ideal for preparing 10 to 100 mL samples prior to chromatography or other instrumental analyses.

Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs.
  • Low protein binding to minimize interaction with your sample and maximize recovery.
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility.

Applications:

  • Buffer Filtration, Clarification of Aqueous Solutions
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Syringe Filter
For Use With (Equipment) Syringe
Material Polypropylene
Material (Membrane) PES
Pore Size 0.45 μm
Sample Volume 100 mL
Sterility Non-sterile
Filtration Area 4.5 cm2
For Use With (Application) Analytical sample prep
Housing Material Polypropylene
Outlet Style Male Luer Slip
Product Line Millex
Quantity 250/Pk.
Type Syringe Filter
Volume (Metric) <200 mL
Diameter (Metric) 33 mm
Diameter (Metric) Filter 33 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.