Learn More
Description
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs.
- Low protein binding to minimize interaction with your sample and maximize recovery.
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility.
Applications:
- Buffer Filtration, Clarification of Aqueous Solutions
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Syringe Filter |
| For Use With (Equipment) | Syringe |
| Material | Polypropylene |
| Material (Membrane) | PES |
| Pore Size | 0.45 μm |
| Sample Volume | 100 mL |
| Sterility | Non-sterile |
| Filtration Area | 4.5 cm2 |
| For Use With (Application) | Analytical sample prep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.