Learn More
MilliporeSigma™ Millex™ Product Container
Catalog No. SLC0NTAIN Shop All MilliporeSigma Products
Change view
Click to view available options
Millex™ Product Container
Catalog No.
SLC0NTAIN Millex™ Product Container
1 options
Catalog No. SLC0NTAIN Supplier MilliporeSigma™ Supplier No. SLC0NTAIN
Add to Cart
Add to Cart
Designed for convenient, benchtop storage of 33 mm Millex syringe filters.
  • Comes with 5 product label cards for convenient, benchtop storage of 33 mm Millex syringe filters
  • Label cards have space to enter both the catalog and lot number for easy identification of the stored filters
  • Holds up to 200 non-sterile 33 mm filters
  • Key applications
    • Aalytical Sample Prep
    • High Performance Liquid Chromatography (HPLC)
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
For Use With (Application) General laboratory use only
Includes Millex™ Product Container with 5 label cards