Learn More
Description
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Low protein binding to minimize interaction with your sample and maximize recovery.
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility.
Applications:
- Buffer Filtration, Clarification of Aqueous Solutions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Connectors | Male Luer Slip |
| Disposable | Single-use |
| Filter Type | Syringe Filter |
| For Use With (Equipment) | Syringe |
| Material | Polypropylene |
| Material (Membrane) | PES |
| Pore Size | 0.22 μm |
| Sample Volume | 100 mL |
| Sterility | Non-sterile |
| Filtration Area | 4.5 cm2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
