Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millex™ Syringe Filter, PES, 0.22 μm
GSA_VA

Catalog No. SLGPX13NL Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric) Filter:
13 mm
33 mm
Quantity:
100/Pk.
1000/Pk.
250/Pk.
50/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Filter Quantity
SLGPX13NL 13 mm 100/Pk.
SLGPX13NK 13 mm 1000/Pk.
SLGP033NB 33 mm 250/Pk.
SLGP033NK 33 mm 1000/Pk.
SLGP033NS 33 mm 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SLGPX13NL Supplier MilliporeSigma™ Supplier No. SLGPX13NL
Add to Cart
Add to Cart

Millex™ PES Syringe Filter, 0.22 μm for removing particulates, reducing signal-to-noise ratios and maintaining flat baselines.

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:

  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Low protein binding to minimize interaction with your sample and maximize recovery.
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility.

Applications:

  • Buffer Filtration, Clarification of Aqueous Solutions

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Code Green
Connectors Male Luer Slip
Disposable Single-use
Filter Type Syringe Filter
For Use With (Equipment) Syringe
Material Polypropylene
Material (Membrane) PES
Pore Size 0.22 μm
Sample Volume 10 mL
Sterility Non-sterile
For Use With (Application) Analytical sample prep
Housing Material Polypropylene
Hydrophobicity Hydrophilic
Inlet Style Male Luer Slip
Outlet Style Male Luer Slip
Quantity 100/Pk.
Type Syringe Filter
Volume (Metric) <10 mL
Diameter (Metric) 13 mm
Diameter (Metric) Filter 13 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.