Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Millicell Hanging Cell Culture Inserts
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
8 μm
No. of Wells:
6
12
24
Color:
Translucent
Transparent
Pore Density:
1 x 108 pores/cm2
2 x 105 pores/cm2
2 x 106 pores/cm2
4 x 106 pores/cm2
Description
- Individually blister-packaged inserts fit in most culture plates
- TC-treated and gamma sterilized
- Pore sizes range from 0.4 to 8.0 μm
- Low pore density membranes are optically transparent
- High pore density membranes are optically translucent
- Applications include co-culture, migration and invasion assays, toxicity testing, transport and permeability assays and organotypic Air-Liquid Interface (ALI) culture

Specifications
Specifications
Culture Area | 4.5 cm2 |
Material | PET |
Volume (Metric) Well | 4 mL |
Volume (Metric) Working | 2 mL |
Pore Size | 8 μm |
Type | Cell Culture Insert |
For Use With (Equipment) | Culture Plate |
Diameter (Metric) Inner | 24 mm |
Diameter (Metric) Outer | 27 mm |
Height (Metric) | 15.5 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction