Learn More
Latex Particles For Millikan
SDPSCI_ED
Catalog No. s52067b
Click to view available options
:
Replacement Latex Spheres
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Requires
Power supply
Replacement Latex Particles for Millikan Apparatus.
  • Use these specially designed latex spheres instead of oil, for an easier and safer way to introduce your students to the Millikan experiment
  • Safer: because the spheres don't require the use of radioactive sources
  • Easier: because the consistent size and reliable changes of droplets limits the calculations required in the experiment
Specifications
Product Type Replacement Latex Spheres
For Use With (Application) Shows the discreteness of an electronic charge and measures the charge on an electron
Includes 0.5 oz bottle of specially designed latex spheres.
Diameter (Metric) 1.02 microns
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.