Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millistak+™ HC Pod Depth Filter

Catalog No. MD0HC01FS1
Click to view available options
Diameter (Metric):
0.11 m2
0.55 m2
This item is not returnable. View return policy
This item is not returnable. View return policy

MilliporeSigma™ Millistak+™ HC Pod Depth Filter features a self-contained and disposable system that protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.

  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Millistak+™ HC dual-action media improves prefiltration and compresses clarification
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces
  • Aqueous extraction contains <0.25 EU/mL as determined by the Limulus Amebocyte Lysate (LAL) clot test technique
  • 1 autoclave cycle of 60 min at 123°C
TRUSTED_SUSTAINABILITY

Specifications

Material Cellulose, Inorganic Filter Aid, Glass Fiber Reinforced Polypropylene
For Use With (Application) Primary clarification directly out of the bioreactor
Grade D0HC
Max. Inlet Pressure 3.5 bar (50 psi) at 25°C; 1 bar (15 psi) at 80°C
Type Millistak+ HC Pod Depth Filter
Length (Metric) 62 cm
Height (Metric) 32 cm
Diameter (Metric) 0.11 m2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.