Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Millon's reagent, for the detection of Tyrosine

Catalog No. ac458541000
Click to view available options
Quantity:
100 mL
500 mL

CAS: 10045-94-0 | HgN2O6 | 324.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10045-94-0
Molecular Formula HgN2O6
Molecular Weight (g/mol) 324.60
MDL Number MFCD00011038
InChI Key ORMNPSYMZOGSSV-UHFFFAOYSA-N
Synonym mercuric nitrate, mercury nitrate, mercury ii nitrate, citrine ointment, mercury dinitrate, millon's reagent, mercury pernitrate, mercury 2+ nitrate, nitrate mercurique french, unii-2fmv9338bw
PubChem CID 24864
IUPAC Name mercury(2+);dinitrate
SMILES [Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Quantity 100 mL
Linear Formula Hg(NO3)2
Formula Weight 324.6
Specific Gravity 1.4
Density 1.4000g/mL
Chemical Name or Material Millon's reagent
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Corrosive t
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 233-152-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.