Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Miltefosine, 98%

Catalog No. ac461830010
Click to view available options
Quantity:
1 g

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58066-85-6
Molecular Formula C21H46NO4P
Molecular Weight (g/mol) 407.57
InChI Key PQLXHQMOHUQAKB-UHFFFAOYSA-N
PubChem CID 3599
ChEBI CHEBI:75283

Specifications

Melting Point 216°C
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 407.57
Percent Purity 98%
Chemical Name or Material Miltefosine
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.