Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mimitin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7328 Supplier Bioss Supplier No. BS6551R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Complex I, or NADH ubiquinone oxidoreductase, is a large protein complex of 950,000 Da molecular weight made up by 45 to 46 different subunits. A total of seven of the subunits of the complex are encoded by mitochondrial DNA, while the remainder subunits are nuclear encoded, which are translated in the cytosol and translocated into the organelle for assembly at the inner membrane. The enzyme complex catalyses electron entry from NADH via a flavin (FMN) and several non-heme iron centers. Complex I is sensitive to a wide range of inhibitors, many of which are pesticides or other common environmental toxins, such as rotenone. Complex I dysfunction is a common cause of genetic OXPHOS defects. Altered functioning of this complex is also thought to contribute to several neurological disorders including Parkinson’s disease and schizophrenia. Also, there is evidence of Complex I involvement in diabetes.

Specifications

Antigen Mimitin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MT-ND2
Gene Accession No. O00217, O00483, O14561, O15239, O43181, O43674, O43676, O43677, O43678, O43920, O75251, O75306, O75380, O75438, O75489, O95139, O95167, O95168, O95169, O95178, O95182, O95298, O95299, O96000, P03886, P03891, P03897, P03901, P03905, P03915, P03923, P17568, P19404, P28331, P49821, P51970, P56181, P56556, Q16718, Q16795, Q86Y39, Q8N183, Q9BU61, Q9NRX3, Q9NX14, Q9P032, Q9P0J0, Q9UI09, Q9Y375, Q9Y6M9
Gene Alias 0610006N12Rik; 0610007D05Rik; 0610010M09Rik; 0610011B04Rik; 0610033L03Rik; 1010001M04Rik; 1010001M12Rik; 1010001N11Rik; 1110002H15Rik; 1110007M04Rik; 1190008J14Rik; 13 kDa differentiation-associated protein; 1300010H20Rik; 13kDa differentiation-associated protein; 14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 14.5kDa; 14kDa; 1500032D16Rik; 1700022J01Rik; 1810004I06Rik; 1810011O01Rik; 1810049F12Rik; 1810058I14Rik; 2010012C24Rik; 2010300P09Rik; 2210401F17Rik; 22kDa; 2310016K22Rik; 2310039H15Rik; 2400007M02Rik; 2410001M24Rik; 2410011G03Rik; 24-kDa subunit of mitochondrial NADH dehydrogenase; 2610003B19Rik; 2700033I16Rik; 2700038D15Rik; 2700054G14Rik; 2900002J19Rik; 2900010C23Rik; 2900010I05Rik; 2900053E13Rik; 2P1; 3000003G13Rik; 4733401H18Rik; 5830412M15Rik; 6720411N02Rik; 8kDa; 9130423F15Rik; 9930026A05Rik; Aa2-258; ACP; Acyl carrier protein, mitochondrial; AGGG; AI256693; AI325567; AI415450; AI987932; AL033311; AQDQ; ASHI; AU015782; AU022060; AV000592; AV006275; AW112974; AW214064; AW261656; B12; B13; B14; B14.5a; B14.5b; B14.7; B15; B16.6; B17; B17.2; B17.2L; B17.2-like; B18; B22; B230217P19Rik; B8; B9; bA22L21.1; BC021616; BC059730; BC064011; BOS_1039; BOS_10761; BOS_11808; BOS_13784; BOS_1388; BOS_15116; BOS_16299; BOS_18159; BOS_1952; BOS_19726; BOS_19776; BOS_19943; BOS_2014; BOS_20464; BOS_20957; BOS_22237; BOS_22436; BOS_22742; BOS_23358; BOS_24411; BOS_24953; BOS_24954; BOS_25118; BOS_25890; BOS_2626; BOS_3615; BOS_3911; BOS_4049; BOS_4411; BOS_4741; BOS_4923; BOS_5279; BOS_6009; BOS_6179; BOS_7096; BOS_7383; BOS_7616; BOS_7617; BOS_8035; BOS_8249; BOS_826; BOS_8620; BOS_9716; C1-18k; C1-B8; C22H3orf60; C3orf60; C6orf66; C86051; C9H6ORF66; CC6; CD14; CDA016; Cell adhesion protein SQM1; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CGI65; CGI-65; CI; CI-10k; CI-13kA; CI-13kB; CI13KDA; CI-13kD-A; CI-13kD-B; CI-15 kDa; CI15K; CI-15k; CI-18; CI-18 kDa; CI-19kD; CI-20; CI-20kD; CI-23k; CI23KD; CI-23kD; CI-24k; CI-30; CI-30kD; CI39k; CI-39k; CI-39kD; CI-42k; CI-42kD; CI-49; CI-49kD; CI-51K; CI51KD; CI-51kD; CI-75k; CI-75kD; CI-9k; CI-9kD; CIA30; CI-AGGG; CI-AQDQ; CI-ASHI; CI-B12; CI-B14; CI-B14.5a; CI-B14.5b; CI-B14.7; CI-B14-5A; CI-B15; CI-B16.6; CI-B17; CIB17.2; CI-B17.2; CI-B18; CI-B22; CIB8; CI-B8; CI-B9; CI-ESSS; CI-KFYI; CI-MLRQ; CI-MNLL; CI-MWFE; CI-PDSW; CI-PGIV; CI-SDAP; CISGDH; CI-SGDH; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 10kDa subunit; complex I 13kDa subunit A; complex I 13kDa subunit B; complex I 18kDa subunit; complex I 20kDa subunit; complex I 23kDa subunit; complex I 24kDa subunit; complex I 30kDa subunit; complex I 39kDa subunit; complex I 42kDa subunit; complex I 49kDa subunit; complex I 51 kda subunit; complex I 51kDa subunit; complex I 75kDa subunit; Complex I 9kDa subunit; complex I AGGG subunit; complex I ASHI subunit; complex I B14 subunit; complex I B14.5a subunit; complex I B14.7 subunit; complex I B15 subunit; complex I B16.6 subunit; complex I B17.2 subunit; complex I B18 subunit; complex I B22 subunit; complex I B8 subunit; complex I B9 subunit; Complex I intermediate-associated protein 30, mitochondrial; complex I KFYI subunit; complex I MNLL subunit; complex I MWFE subunit; complex I NP17.3 subunit; complex I PDSW subunit; complex I SDAP subunit; complex I SGDH subunit; Complex I subunit B13; complex I subunit B14.5b; complex I, mitochondrial respiratory chain; complex I, mitochondrial respiratory chain, 10-kD subunit; complex I, mitochondrial respiratory chain, 13-kD subunit; complex I, mitochondrial respiratory chain, 20-KD subunit; complex I, mitochondrial respiratory chain, 75-kD subunit; complex I, mitochondrial respiratory chain, B17 subunit; complex I, mitochondrial respitory chain, 24 kD subunit; Complex I-13kD-A; Complex I-13kD-B; Complex I-15 kDa; Complex I-18 kDa; Complex I-19kD; Complex I-20kD; Complex I-23kD; Complex I-30kD; Complex I-39kD; Complex I-42kD; Complex I-49kD; Complex I-51kD; Complex I-7
Gene Symbols MT-ND1, MT-ND2, MT-ND3, MT-ND4, MT-ND4L, MT-ND5, MT-ND6, NDUFA1, NDUFA10, NDUFA11, NDUFA12, NDUFA13, NDUFA2, NDUFA3, NDUFA4, NDUFA4L2, NDUFA5, NDUFA6, NDUFA7, NDUFA8, NDUFA9, NDUFAB1, NDUFAF1, NDUFAF2, NDUFAF3, NDUFAF4, NDUFB1, NDUFB10, NDUFB11, NDUFB2, NDUFB3, NDUFB4, NDUFB5, NDUFB6, NDUFB7, NDUFB8, NDUFB9, NDUFC1, NDUFC2, NDUFS1, NDUFS2, NDUFS3, NDUFS4, NDUFS5, NDUFS6, NDUFS7, NDUFS8, NDUFV1, NDUFV2, NDUFV3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Mimitin .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126328, 25915, 29078, 374291, 4535, 4536, 4537, 4538, 4539, 4540, 4541, 4694, 4695, 4696, 4697, 4698, 4700, 4701, 4702, 4704, 4705, 4706, 4707, 4708, 4709, 4710, 4711, 4712, 4713, 4714, 4715, 4716, 4717, 4718, 4719, 4720, 4722, 4723, 4724, 4725, 4726, 4728, 4729, 4731, 51079, 51103, 54539, 55967, 56901, 91942
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.