Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINA Antibody (1E8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | MINA |
---|---|
Clone | 1E8 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
MINA Monoclonal antibody specifically detects MINA in Human samples. It is validated for ELISASpecifications
MINA | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52 | |
A synthesized peptide derived from Human MINA [UniProt Q8IUF8] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1E8 | |
ELISA | |
Unconjugated | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
84864 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title