Learn More
Invitrogen™ MINA53 Monoclonal Antibody (M532)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 397300

Description
MINA53 Monoclonal Antibody for Western Blot, ICC/IF, IHC, ELISA
MINA is nuclear localized, myc-inducible protein that is thought to play a role in mammalian cell proliferation. Treatment of cancer cells lines such as the colon cancer cell line SW680 with siRNA against MINA inhibits cell growth, demonstrating that MINA may be a potential therapeutic target. MINA regulates several genes related to cell adhesion and metabolism that have also been shown to be regulated by c-Myc, but also regulates other genes whose expression are not modulated by c-Myc such as EGFR, IL-6 and HGF. MINA has also been found to act as a repressor to IL-4 expression in T cells, indicating that it may also play a role in T cell differentiation and genetic variation in T helper type 2 bias.
Specifications
MINA53 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q8IUF8 | |
RIOX2 | |
Recombinant full-length human Mina53. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
M532 | |
Unconjugated | |
RIOX2 | |
1810047J07Rik; 2410057H13Rik; 3830408E23Rik; 60S ribosomal protein L27a histidine hydroxylase; AI449204; Bifunctional lysine-specific demethylase and histidyl-hydroxylase MINA; DKFZp762O1912; FLJ14393; histone lysine demethylase MINA; MDIG; mina; mina {ECO:0000312; Mina53; Mina-53; mineral dust induced gene protein; mineral dust-induced gene protein; MYC induced nuclear antigen; myc-induced nuclear antigen; myc-induced nuclear antigen, 53 kDa; NO52; Nucleolar protein 52; Protein mina53; RGD:708521}; Ribosomal oxygenase 2; Ribosomal oxygenase MINA; RIOX2; ROX | |
Mouse | |
Protein A | |
RUO | |
84864 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.