Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mind Bomb 2/MIB2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068224V
Description
Mind Bomb 2/MIB2 Polyclonal antibody specifically detects Mind Bomb 2/MIB2 in Human samples. It is validated for Western BlotSpecifications
Mind Bomb 2/MIB2 | |
Polyclonal | |
Western Blot | |
E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, Mind bomb homolog 2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1 | |
The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human mindbomb homolog 2 (Skeletrophin) using the numbering given in entry NP_543151.1 (GeneID 142678). | |
0.1 mL | |
Signal Transduction | |
142678 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction