Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral oil

Catalog No. AA31911D9
Click to view available options
Quantity:
1 kg
5 x 1 kg

Catalog No. AA31911D9

Supplier: Thermo Scientific Chemicals 031911D9

Only null left
Add to Cart

CAS: 8020-83-5

Mineral oil is used as an overlay covering microdrops of culture medium in petri dishes. It is an active ingredient in baby lotions, cold creams, ointments and cosmetics. It is also used in electrical and mechanical industries as thermal fluid in electrical components and non -conductive coolant. It finds application as a transformer oil, as an anti rust agent for blades and as an insulator in high voltage switchgear. Further, it is used to store reactive metals like lithium, sodium, etc and in X-ray crystallography to suspend crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8020-83-5
MDL Number MFCD00131611

Specifications

Color Colorless
Flash Point >100°C (212°F)
Odor Mineral-oil-like
Quantity 5 x 1 kg
Solubility Information Miscible with ether. Slightly miscible with ethanol. Immiscible with water.
Physical Form Liquid
Chemical Name or Material Mineral oil
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.