Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral oil, high purity

Catalog No. p-7042457
Click to view available options
Quantity:
100 mL
500 mL

Suitable for PCR applications | CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

White Mineral Oils find use in a wide variety of industries and applications including personal care products, cosmetics, pharmaceuticals, food processing and plastics. It can be used as a plasticizer, catalyst carrier and pigment dispersant. Mineral oils are used in several mechanical and industrial capacities as thermal fluid or non-conductive coolants in electric components because they do not conduct electricity but do displace air and water. It is used in medicinal or cosmetic applications or food processing and fulfils stringent purity requirements of different pharmacopoeias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
White Mineral Oils find use in a wide variety of industries and applications including personal care products, cosmetics, pharmaceuticals, food processing and plastics. It can be used as a plasticizer, catalyst carrier and pigment dispersant. Mineral oils are used in several mechanical and industrial capacities as thermal fluid or non-conductive coolants in electric components because they do not conduct electricity but do displace air and water. It is used in medicinal or cosmetic applications or food processing and fulfils stringent purity requirements of different pharmacopoeias.

Solubility
It is insoluble in water. Soluble in ether. Soluble in chloroform, petroleum spirit, ligroin, carbondisulfide, glycerol, oil .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Flash Point >110°C (230°F)
Odor Odorless
Quantity 100 mL
Solubility Information It is insoluble in water. Soluble in ether. Soluble in chloroform,petroleum spirit,ligroin,carbondisulfide,glycerol,oil .
Physical Form Oily Liquid
Chemical Name or Material Mineral oil
Grade High purity
EINECSNumber 232-455-8
RTECSNumber PY8047000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.