Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Oil, Medium, Reagents

Catalog No. 01337167
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-167 500 mL
01-337-170 1 L
01-337-168 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-167 Supplier Reagents Holdings Llc Supplier No. C2251700500C

Mineral Oil, Medium, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 452.363

Specifications

Boiling Range >230°C
Color Colorless
Boiling Point >230°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 0.85
Grade Reagent
Physical Form Liquid
Chemical Name or Material Mineral Oil, Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.