Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mineralocorticoid R/NR3C2 Antibody (H10E4C9F), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300562MFV610
Description
Mineralocorticoid R/NR3C2 Monoclonal antibody specifically detects Mineralocorticoid R/NR3C2 in Human, Mouse, Rat, Chicken, Rabbit, Sheep samples. It is validated for Flow CytometrySpecifications
Mineralocorticoid R/NR3C2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
4306 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
H10E4C9F | |
Flow Cytometry | |
FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
Aldosterone-3 | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Chicken, Rabbit, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction