Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mineralocorticoid R/NR3C2 Antibody (H10E4C9F), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300562PE
Description
Mineralocorticoid R/NR3C2 Monoclonal specifically detects Mineralocorticoid R/NR3C2 in Human, Mouse, Rat, Chicken, Rabbit, Sheep samples. It is validated for Flow Cytometry.Specifications
Mineralocorticoid R/NR3C2 | |
Monoclonal | |
PE | |
P08235 | |
NR3C2 | |
Aldosterone-3 | |
0.1 mL | |
Primary | |
Detects mineralocorticoid receptor (MR). | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
H10E4C9F | |
Flow Cytometry | |
FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
Mouse | |
Protein A purified | |
RUO | |
4306 | |
Human, Mouse, Rat, Chicken, Rabbit, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction