Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mineralocorticoid R/NR3C2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Mineralocorticoid R/NR3C2 |
---|---|
Dilution | Western Blot 1:500-1:2000, Simple Western, Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
Applications | Western Blot, Simple Western, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Mineralocorticoid R/NR3C2 Polyclonal antibody specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
Mineralocorticoid R/NR3C2 | |
Western Blot, Simple Western, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Mineralocorticoid R/NR3C2. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:2000, Simple Western, Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
Polyclonal | |
Purified | |
RUO | |
0.2 μm filtered solution in PBS | |
4306 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title