Learn More
American Scientific Mini Ripple Tank
SDPSCI_ED
Catalog No. S13181
Click to view available options
:
Quantity: 1
Includes instructions, 3 Acrylic shapes for refraction, 3 Dippers for plane and circular, and power adapter
  • This apparatus uses modern technology to completely update the Ripple Tank concept
  • It provides a compact and elegant method of showing the wave phenomena of diffraction, refraction, reflection, and interference, with none of the usual problems
  • The tank is a 4 in. (100 mm) square and is supported on a base which provides the wave generator and lighting
  • Images of the water waves produced are projected upwards onto a frosted screen, which is viewed from above
  • The white light source can be switched between constant illumination and strobe to give either moving or frozen images on the screen
  • In the strobe mode, the light source is synchronized to the wave generator so that perfectly static patterns are displayed
  • The wave frequency can be varied, giving wavelengths in the tank ranging from several cm to a few mm
Specifications
For Use With (Application) Ripple Tank concept demonstration
Quantity 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.