Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Mini Spatula Extension Handle

Catalog No. 18100976 Shop All Excelta Corporation Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-100-976 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-100-976 Supplier Excelta™ Supplier No. 260H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Excelta™ 260H extension handle is for use with the 260 series mini spatulas.

  • The 260H three star extension handle for the 260 series mini-spatulas are made of polished stainless steel.
  • They are hollow allowing them to fit over any of our 260 spatulas.
  • The extended handle facilitates the use of the 260 series and ergonomically enhances the tool.

Specifications

Color White
Disposable Reusable
Product Type Miniature Spatula Extension Handle
For Use With (Equipment) Mini Spatulas, Oilers
Length (English) 2.75 in.
Blade Material Stainless Steel, Plastic
Material Stainless Steel (440)
Length (Metric) 68.75 mm
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.