Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

minicore HCV Mouse anti-Virus, Unconjugated, Clone: Neo4, MilliporeSigma™

Catalog No. MABF2057100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2057-100 100 μg
MABF2057-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2057-100 Supplier MilliporeSigma Supplier No. MABF2057100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-minicore HCV, clone Neo4, Cat. No. MABF2057, is a mouse monoclonal antibody that detects intracellular and secreted HCV core and minicore proteins in HCV infected cells and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen minicore HCV
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone Neo4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias HCV;Hepatitis C virus minocores
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal region of Hepatitis C virus (HCV) core protein.
Molecular Weight of Antigen ∽ 8 and 21 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone Neo4 detects HCV core and minicore proteins in cells infected with HCV virus.
Target Species Virus
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.