Learn More
Minigrip™ GreenLine™ Biodegradable Reclosable Zipper Bags
Catalog No. p-4676244
Click to view available options
Dimensions (L x W):
10.16 x 15.2cm (4 x 6 in.)
12.7 x 17.7cm (5 x 7 in.)
15.2 x 22.8cm (6 x 9 in.)
22.8 x 30.4cm (9 x 12 in.)
5.08 x 5.08cm (2 x 2 in.)
5.08 x 7.6cm (2 x 3 in.)
7.06 x 10.16cm (3 x 4 in.)
For use with all kinds of lab, medical and food storage
  • Made of low-density polyethylene
  • Thickness of 2 mil
  • Bags tested to biodegrade 15% within 12 months per ASTM D5511 test results when in contact with other materials that were also biodegrading
  • Available in variety of sizes
Compliance
Proven: ASTM D5511 tested by an independent lab to be biodegradable
Specifications
Thickness (English) 2 mil
Dimensions (L x W) 5.08 x 5.08cm (2 x 2 in.)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.