Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Lab Guard™ Specimen Bags with TearZone™ and Destroyable Biohazard Symbol

Catalog No. 22311107
Click to view available options
Color:
Green
Red
Red Tint
Yellow

Safe and durable specimen-handling reclosable bags feature open and close zipper technology and grip edges

With TearZone™ destroyable biohazard symbol feature.

  • Convenient 6 by 9 in. size
  • Color codes can be used to identify a specific urgency or department
  • TearZone™ feature will not leak — contains leak to one bag for overall cleanup and savings
  • Tears open easily at processing — no need for sharps to open the bag quickly
  • Grip edges make opening easier with gloved hands
  • Vivid biohazard risk warning graphic
  • Separate pouch for paperwork storage
  • Option to dispose unsoiled bags in regular waste means additional cost saving

Specifications

Color Red
For Use With (Application) Biohazard specimen transport
Includes TearZone™ and destroyable biohazard symbol
Material Virgin Polyethylene
Thickness (Metric) 0.279 mm
Thickness (English) 2 mil
Dimensions (L x W) 9 x 6 in. (22.8 x 15.2 cm)
Length (English) 9 in.
Length (Metric) 22.8 cm
Width (English) 6 in.
Width (Metric) 15.2 cm
Product Type Biohazard Specimen Transport Bag
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.