Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ LabGuard™ Chemotherapy Bags with Absorbent Pad

Catalog No. p-4654101
Click to view available options
Height (Metric):
30.48 cm
38.10 cm
Width (Metric):
22.86 cm
30.48 cm

Used for safe transport of chemotherapy drugs

Doctors, hospitals, and pharmacies used these secondary storage containers to transport specimen containers from the pharmacy to the patient and return of the empty containers to the pharmacy.

  • With reclosable zipper
  • Thickness of 4 mil
  • Available in two standard sizes

Specifications

Closure Type Ziplock
Color Clear
For Use With (Application) Safe transport of chemotherapy drugs from the pharmacy to the patient and return of the empty containers to the pharmacy
Includes Absorbent pad, reclosable zipper, yellow and black chemotherapy print
Material Plastic
Quantity 500 Case
Thickness (English) 4 mil
Width (Metric) 30.48 cm
Height (Metric) 38.10 cm
Product Type Chemotherapy Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.