Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Minigrip™ Slider Grip™ Storage Bags
Click to view available options
Dimensions (L x W):
20.32 x 17.78cm
22.86 x 30.48cm
26.67 x 27.94cm
38.83 x 40.64cm
Description
- Made from sturdy polyethylene
- Thickness of 2.7 mil
- Available in quart, letter, 1 and 2 gal. sizes
- Reduces unnecessary repetitive motions
Specifications
Specifications
| Closure Type | Zip Seal |
| Color | Clear with Green Zipper |
| Material | Polyethylene |
| Dimensions (L x W) | 38.83 x 40.64cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction