Learn More
Minigrip™ StaticGuard™ Anti-Static Bags
Description
- Nonbiohazard
- With zipper
- Thickness of 4 mil
- Pink
- Available in different sizes
Specifications
Specifications
| Description | Anti-Static Bag |
| Closure Type | Zip Seal |
| Color | Pink |
| Quantity | 500/Pk. |
| Thickness (English) | 4 mil |
| Dimensions (L x W) | 13 x 18 in. (33.02 x 45.72cm) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.