Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ UV Guard™ UV Protection Bags

Catalog No. 22311213
Click to view available options
Dimensions (L x W):
12 x 12 in. (30.48 x 30.48cm)
2 x 3 in. (5.08 x 7.62cm)
2.5 x 9 in. (6.35 x 22.86cm)
3 x 5 in. (7.62 x 12.70cm)
4 x 6 in. (10.16 x 15.24cm)
5 x 8 in. (12.7 x 20.32cm)
6 x 8 in. (15.24 x 20.32cm)
8 x 14 in. (20.32 x 35.56cm)
8 x 8 in. (20.32 x 20.32cm)

Catalog No. 22-311-213

Supplier: Minigrip™ MGUV3P0808

Only null left
Add to Cart
Edge
Add to Cart

Prevent light-sensitive products from deterioration

  • Amber
  • With Color Line™ and premium zipper
  • Film contains UV blocker
  • Thickness of 3 mil
  • Available in different sizes

Compliance

USP XXII and USP XXIII procedure 66.1.

Specifications

Closure Type Premium Zipper
Color Amber
Material Polyethylene
Thickness (English) 3 mil
Dimensions (L x W) 8 x 8 in. (20.32 x 20.32cm)
Length (English) 8 in.
Width (English) 8 in.
Product Type UV Protection Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.