Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MiniMark™ Industrial Label Printer Ribbons

Catalog No. p-7070671
Click to view available options
Color:
Black
Blue
Green
Orange
Red
White
Yellow

Create affordable custom identification with the MiniMark industrial label printer. Brady™ MiniMark™ Industrial Label Printer Ribbons are available in a variety of colors.

  • Pre-loaded onto spindles with "keyed" end caps
  • Tough, smear proof ribbons produce long-lasting printed text
  • For industrial and outdoor areas

Specifications

Product Type MiniMark Print Ribbons
Color Yellow
For Use With (Equipment) MiniMark Printer
Length (English) 290 ft.
Dimensions (L x W) 290 ft. x 4 in.
Quantity 2/Pk.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.