Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ MiniPet™ Pipette for Cholesterol Analyzers

Catalog No. p-4881409
Click to view available options
Capacity (Metric):
40 μL
Sample Type:
Blood

For use with the Cholestech LDX™ System.

For use with Cholestech LDX lipid monitoring system.

For use with Cholestech LDX lipid monitoring system.

Specifications

Type MiniPet™ Pipette
Sample Type Blood
For Use With (Equipment) Cholestech LDX™ lipid monitoring system
Capacity (Metric) 40 μL
Content And Storage 15°C to 30°C (59°F to 86°F)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.