Learn More
Promega MINK1 Kinase Enzyme System, 1mg
Catalog No. PRV3912 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3912 1 mg
1 options
Catalog No. PRV3912 Supplier Promega Supplier No. V3912
Add to Cart
Add to Cart
Request a Quote
Recombinant human MINK1 (1-320) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. MINK1 is a member of the germinal center family of kinases that are homologous to the Ste20 family and regulate a wide variety of cellular processes.
  • Easily Screen and Profile MINK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C