Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MINK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528655 Supplier Invitrogen™ Supplier No. PA528655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (91%), Xenopus laevis (94%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MINK1 is a serine/threonine kinase belonging to the germinal center kinase (GCK) family. The protein is structurally similar to the kinases that are related to NIK and may belong to a distinct subfamily of NIK-related kinases within the GCK family. Studies of the mouse homolog indicate an up-regulation of expression in the course of postnatal mouse cerebral development and activation of the cJun N-terminal kinase (JNK) and the p38 pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MINK1
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MINK1
Gene Accession No. Q8N4C8
Gene Alias B55; FLJ18426; FLJ38123; FLJ94103; GCK family kinase MiNK; hMINK; hMINKbeta; ISK; JLNS; JLNS2; LQT2/5; LQT5; Map4k6; MAPK/ERK kinase kinase kinase 6; MEK kinase kinase 6; MEKKK 6; MGC33114; MINK; Mink 1; Mink 2; MINK1; misshapen like kinase 1; misshapen/NIK-related kinase; Misshapen/NIKs-related kinase; misshapen-like kinase 1; misshapen-like kinase 1 (zebrafish); mitogen-activated protein kinase kinase kinase kinase 6; Yeast Sps1/Ste20-related kinase 2; Ysk2; ZC3
Gene Symbols MINK1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human MAP4K6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.