Learn More
Invitrogen™ MINK1 Polyclonal Antibody

Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (87%), Xenopus laevis (89%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MINK1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | MINK1 |
| Gene Accession No. | Q8N4C8 |
| Gene Alias | B55; FLJ18426; FLJ38123; FLJ94103; GCK family kinase MiNK; hMINK; hMINKbeta; ISK; JLNS; JLNS2; LQT2/5; LQT5; Map4k6; MAPK/ERK kinase kinase kinase 6; MEK kinase kinase 6; MEKKK 6; MGC33114; MINK; Mink 1; Mink 2; MINK1; misshapen like kinase 1; misshapen/NIK-related kinase; Misshapen/NIKs-related kinase; misshapen-like kinase 1; misshapen-like kinase 1 (zebrafish); mitogen-activated protein kinase kinase kinase kinase 6; Yeast Sps1/Ste20-related kinase 2; Ysk2; ZC3 |
| Gene Symbols | MINK1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.