Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINOS1 Antibody (OTI4H7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272692APC
Description
MINOS1 Monoclonal antibody specifically detects MINOS1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MINOS1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
440574 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI4H7 | |
Western Blot, Immunohistochemistry | |
mitochondrial inner membrane organizing system 1 | |
Full length human recombinant protein of human C1orf151(NP_001007535) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction