Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINOS1 Antibody (OTI4H7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272692UV
Description
MINOS1 Monoclonal specifically detects MINOS1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MINOS1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MINOS1 | |
Full length recombinant protein of human MINOS1 (NP_001007535) produced in HEK293T cells. | |
0.1 mL | |
440574.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4H7 | |
Western Blot, Immunohistochemistry | |
mitochondrial inner membrane organizing system 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction