Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINOS1 Antibody (OTI4H7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245550
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
MINOS1 Monoclonal specifically detects MINOS1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MINOS1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
mitochondrial inner membrane organizing system 1 | |
Mouse | |
8.6 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
OTI4H7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
MINOS1 | |
Full length human recombinant protein of human C1orf151(NP_001007535) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
440574 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction