Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minoxidil, Powder, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18604638
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail

M1138, 38304-91-5, C9H15N5O

Spectrum™ Chemical Minoxidil, Powder, USP is an antihypertensive vasodilator used to help hair growth in the treatment of male pattern baldness. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 38304-91-5
Molecular Formula C9H15N5O
Molecular Weight (g/mol) 209.25
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
IUPAC Name 6-amino-2-imino-4-(piperidin-1-yl)-1,2-dihydropyrimidin-1-ol
SMILES NC1=CC(=NC(=N)N1O)N1CCCCC1

Specifications

CAS Max % 1
Ignition Residue 0.005
Loss on Drying 0.005
Packaging Poly Pail
Quantity 1 kg
Percent Purity 97 to 103%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.