Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINPP1 Antibody (OTI1B2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272693
Description
MINPP1 Monoclonal specifically detects MINPP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MINPP1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:4000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 3.1.3.62, HIPER1, Inositol (13,4,5)-tetrakisphosphate 3-phosphatase, Ins(13,4,5)P(4) 3-phosphatase, MINPP2, MIPPDKFZp564L2016, multiple inositol polyphosphate histidine phosphatase, 1, multiple inositol polyphosphate phosphatase 1, multiple inositol polyphosphate phosphatase 2, multiple inositol-polyphosphate phosphatase 1 | |
Mouse | |
54.9 kDa | |
100 μg | |
9562 | |
Human | |
Lyophilized |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1B2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MINPP1 | |
Full length human recombinant protein of human MINPP1(NP_004888) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction