Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MINPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577263 Supplier Invitrogen™ Supplier No. PA577263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

MINPP1 encodes multiple inositol polyphosphate phosphatase; an enzyme that removes 3-phosphate from inositol phosphate substrates. It is the only enzyme known to hydrolzye inositol pentakisphosphate and inositol hexakisphosphate. This enzyme also converts 2,3 bisphosphoglycerate (2,3-BPG) to 2-phosphoglycerate; an activity formerly thought to be exclusive to 2,3-BPG synthase/2-phosphatase (BPGM) in the Rapoport-Luebering shunt of the glycolytic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MINPP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene MINPP1
Gene Accession No. O35217, Q9UNW1, Q9Z2L6
Gene Alias 2,3-bisphosphoglycerate 3-phosphatase; 2,3-BPG phosphatase; AA408516; HIPER1; Inositol (1,3,4,5)-tetrakisphosphate 3-phosphatase; ins(1,3,4,5)P(4) 3-phosphatase; MINP1; MINPP1; MINPP2; MIPP; Mipp1; MNCb-1572; multiple inositol polyphosphate histidine phosphatase 1; multiple inositol polyphosphate histidine phosphatase, 1; multiple inositol polyphosphate phosphatase 1; multiple inositol polyphosphate phosphatase 2; multiple inositol-polyphosphate phosphatase 1; RP11-57C13.1; UNQ900/PRO1917
Gene Symbols MINPP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 330-382 of Human MIPP.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17330, 29688, 9562
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.