Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Minretumomab Chimeric Recombinant Rabbit Monoclonal Antibody (Minretumomab (CC49))
GREENER_CHOICE

Catalog No. PIMA548179
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548179 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548179 Supplier Invitrogen™ Supplier No. MA548179
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody is specific for the disaccharide sialyl Tn epitope of human TAG-72. TAG-72 is expressed on a wide range of human carcinomas, including colorectal cancers.

Minretumomab is a biosimilar that targets TAG-72.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Minretumomab Chimeric
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone Minretumomab (CC49)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Il5
Gene Accession No. P05113
Gene Alias Mab CC-49
Host Species Rabbit
Immunogen This antibody was raised by immunising BALB/c mice with TAG-72 purified from LS-174T human carcinoma xenograft.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.