Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MINT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536573 Supplier Invitrogen™ Supplier No. PA536573
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 61 kDa. Purity is >95% by SDS-PAGE.

The munc-18 interacting protein (Mint) protein family is a group of evolutionarily conserved adaptor proteins that function in membrane transport and organization. In mammals, there exist three mint isoforms, Mint1, 2, and 3. Although there is little amino acid sequence conservation in the amino-terminal half, the carboxy-terminal half of these proteins is highly conserved. Within this conserved portion there exists a phosphotyrosine-binding (PTB) and a PSD-95/DLG-A/ZO-1 (PDZ) domain, which function as protein interaction modules. Mint1 and 2 appear to be expressed exclusively in the brain and are found to bind to Munc18, an essential component of the synaptic vesicle fusion machinery. Mint3 is ubiquitously expressed in all tissues and is expressed at the lowest levels in the brain and testis. Studies show that mint3 does not interact with munc-18. Mint3 has been found to interact with the Alzheimer's Disease-related amyloid precursor protein (APP) and does so through its PTB and PDZ domains. It has been suggested that mint3 links APP to other transport machinery components, thereby regulating it transport, endocytosis, and metabolism. Abnormal APP metabolism has been shown to be the cause of an early-onset type of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MINT3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene APBA3
Gene Accession No. O96018
Gene Alias Adapter protein X11gamma; amyloid beta (A4) precursor protein-binding family A member 3 (X11-like 2); amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2); amyloid beta (A4) precursor protein-binding, family A, member 3; amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2); amyloid beta A4 precursor protein-binding family A member 3; amyloid beta precursor protein binding family A member 3; Amyloid-beta A4 precursor protein-binding family A member 3; Apba3; AW208791; lin 10; lin-10; MGC:15815; Mint 3; MINT3; mint-3; Neuronal Munc18-1-interacting protein 3; neuron-specific X11L2 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; X11gamma; X11L2; X11-like 2 protein
Gene Symbols APBA3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 371-420 of human X11 gamma.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.