Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIOS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560162 Supplier Invitrogen™ Supplier No. PA560162
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACKFLSDTQL NRYIEKLTNE MKEAGNLEGI LLTGLTKDGV DLMESYVDRT GDVQTASYCM LQGSPLDVLK DERVQYWIEN YRNLLDAWRF WHKRAEFDIH RSKLDPSSKP LAQVFVSCNF CG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

As a component of the GATOR subcomplex GATOR2, functions within the amino acid-sensing branch of the TORC1 signaling pathway. Indirectly activates mTORC1 and the TORC1 signaling pathway through the inhibition of the GATOR1 subcomplex (PubMed:23723238). It is negatively regulated by the upstream amino acid sensors SESN2 and CASTOR1 (PubMed:25457612, PubMed:27487210). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIOS
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIOS
Gene Accession No. Q9NXC5
Gene Alias C81488; FLJ20323; GATOR complex protein MIOS; H_DJ1159O04.1; meiosis regulator for oocyte development; MIO; Mios; Missing oocyte meiosis regulator homolog; missing oocyte, meiosis regulator, homolog; missing oocyte, meiosis regulator, homolog (Drosophila); RGD1308432; Sea4; WD repeat-containing protein mio; WUGSC:H_DJ1159O04.1
Gene Symbols MIOS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MIOS. Recombinant protein control fragment (Product #RP-92791).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54468
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.