Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIOX Antibody (OTI4G7), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272697APCCY7
Description
MIOX Monoclonal specifically detects MIOX in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
MIOX | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
MIOX | |
Full length recombinant protein of human MIOX (NP_060054) produced in HEK293T cells. | |
0.1 mL | |
55586.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI4G7 | |
Flow Cytometry | |
aldehyde reductase (aldose reductase) like 6, Aldehyde reductase-like 6, ALDRL6, inositol oxygenase, Kidney-specific protein 32, KSP32, MGC90217, MI oxygenase, myo-inositol oxygenaseEC 1.13.99.1, Renal-specific oxidoreductase, RSOR | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction