Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558840
Description
Immunogen sequence: QWAVVGDTFP VGCRPQASVV FCDSTFQDNP DLQDPRYSTE LGMYQPHCGL DRVLMSWGHD EYMYQVMKFN KFSL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
MIOX (Myo-Inositol Oxygenase) is a Protein Coding gene. Among its related pathways are superpathway of inositol phosphate compounds and Porphyrin and chlorophyll metabolism. Gene Ontology (GO) annotations related to this gene include iron ion binding and oxidoreductase activity, acting on NAD(P)H.
Specifications
| MIOX | |
| Polyclonal | |
| Unconjugated | |
| MIOX | |
| 0610009I10Rik; AI314022; aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; aldehyde reductase-like 6; Aldrl6; C85427; Inositol oxygenase; kidney-specific protein 32; KSP32; MI oxygenase; Miox; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxidoreductase; renal-specific oxido-reductase; Rsor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGB7 | |
| MIOX | |
| Recombinant protein corresponding to Human MIOX. Recombinant protein control fragment (Product #RP-95354). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction