Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIP-1 alpha Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990250
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0250 100 μL
50-199-0251 25 μL
50-199-0249 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-0250 Supplier Rockland Immunochemicals Supplier No. 1094013150100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. In ELISA formats and other immunoreactive assays, this antibody will recognize recombinant and native human MIP-1a present in body fluids and cell supernatants. This antiserum has not been evaluated for its ability to stain human MIP-1a in tissue sections, nor for its ability to neutralize human MIP-1a in bioassays, nor for its performance in immunoblot analysis.

Specifications

Antigen MIP-1 alpha
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.01% sodium azide; pH
Host Species Rabbit
Immunogen The whole rabbit serum was prepared by repeated immunizations with recombinant human MIP-1a produced in E.coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.